1-Iodo-4,4,4-trifluorobutane 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane
PC7385-5G PC7385-25G
APOSPC7385-5GEA 94.4 EUR
APOSPC7385-5G APOSPC7385-25G
1-Iodo-4,4,4-trifluorobutane 99%
1-Iodo-4,4,4-trifluorobutane
Formula: C₄H₆F₃I
MW: 237.99 g/mol
Boiling Pt: 126…127 °C
Density: 1.85 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00041543
CAS Number: 461-17-6