1,1,1-Trifluoro-2-propanol 97%
Supplier: Apollo Scientific
Danger
PC7386-100G PC7386-25G
APOSPC7386-100GEA 574 EUR
APOSPC7386-100G APOSPC7386-25G
1,1,1-Trifluoro-2-propanol 97%
1,1,1-Trifluoro-2-propanol
Formula: C₃H₅F₃O
Boiling Pt: 75…76 °C
Density: 1.259 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00014403
CAS Number: 374-01-6
EINECS: 206-773-2
UN: 1987
ADR: 3,II