5,5,5-Trifluoro-DL-leucine 97%
Supplier: Apollo Scientific
|
Warning
|
PC7392-1G
PC7392-250MG
PC7392-100MG
APOSPC7392-1GEA
1540
EUR
APOSPC7392-1G
APOSPC7392-250MG
APOSPC7392-100MG
5,5,5-Trifluoro-DL-leucine 97%
5,5,5-Trifluoro-DL-leucine
Formula:
C₆H₁₀F₃NO₂ MW: 185.15 g/mol Melting Pt: 272…275 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039528 CAS Number: 2792-72-5 |