2-Bromo-4-fluoro-1-iodobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-1-fluoro-4-iodobenzene, 2-Bromo-4-fluoroiodobenzene, 1-Bromo-5-fluoro-2-iodobenzene
PC7393-100G PC7393-25G PC7393-500G
APOSPC7393-100GEA 66.7 EUR
APOSPC7393-100G APOSPC7393-25G APOSPC7393-500G
2-Bromo-4-fluoro-1-iodobenzene 98%
2-Bromo-4-fluoro-1-iodobenzene
Formula: C₆H₃BrFI
Boiling Pt: 224…225 °C
Density: 2.323 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00142868
CAS Number: 202865-73-4