2-Bromo-4-fluoro-6-methylaniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-4-fluoro-o-toluidine
PC7404-1G PC7404-5G PC7404-10G
APOSPC7404-1GEA 21.9 EUR
APOSPC7404-1G APOSPC7404-5G APOSPC7404-10G
2-Bromo-4-fluoro-6-methylaniline 98%
2-Bromo-4-fluoro-6-methylaniline
Formula: C₇H₇BrFN
MW: 204.04 g/mol
Melting Pt: 32…36 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01861196
CAS Number: 202865-77-8