3'-Trifluoromethoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3'-(Trifluoromethoxy)acetophenone, 1-[3-(Trifluoromethoxy)phenyl]-1-ethanone, 1-Ethanoyl-3-(trifluormethoxy)benzol
PC7433D-1G PC7433D-25G PC7433D-5G
APOSPC7433D-1GEA 21.9 EUR
APOSPC7433D-1G APOSPC7433D-25G APOSPC7433D-5G
3'-Trifluoromethoxyacetophenone 98%
3'-Trifluoromethoxyacetophenone
Formula: C₉H₇F₃O₂
MW: 204.15
Boiling Pt: 55 °C/5mmHg
Density: 1.285 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00042403
CAS Number: 170141-63-6