2-(Trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
|
Warning
|
PC7433Y-25G
PC7433Y-1KG
APOSPC7433Y-25GEA
0
EUR
APOSPC7433Y-25G
APOSPC7433Y-1KG
2-(Trifluoromethoxy)aniline 98%
2-(Trifluoromethoxy)aniline
Formula:
C₇H₆F₃NO Boiling Pt: 61…63 °C/15mmHg Density: 1.301 g/cm³ (25 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035959 CAS Number: 1535-75-7 EINECS: 216-257-9 UN: 1993 ADR: 3,III |