3-(Trifluoromethoxy)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC7434F-25G PC7434F-5G PC7434F-100G
APOSPC7434F-25GEA 52.2 EUR
APOSPC7434F-25G APOSPC7434F-5G APOSPC7434F-100G
3-(Trifluoromethoxy)benzaldehyde 97%
3-(Trifluoromethoxy)benzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 83…86 °C/24mmHg
Melting Pt: 450
Density: 1.33 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00042406
CAS Number: 52771-21-8