2-(Trifluoromethoxy)benzamide 97%
Supplier: Apollo Scientific
PC7434P-25G PC7434P-5G PC7434P-1G
APOSPC7434P-25GEA 237 EUR
APOSPC7434P-25G APOSPC7434P-5G APOSPC7434P-1G
2-(Trifluoromethoxy)benzamide 97%
2-(Trifluoromethoxy)benzamide
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00042407
CAS Number: 127979-74-2