4-(Trifluoromethoxy)benzamidoxime 97%
Supplier: Apollo Scientific
Warning
Synonyms: N-Hydroxy-4-(trifluoromethoxy)benzimidamide
PC7436D-25G PC7436D-5G PC7436D-1G
APOSPC7436D-25GEA 203 EUR
APOSPC7436D-25G APOSPC7436D-5G APOSPC7436D-1G
4-(Trifluoromethoxy)benzamidoxime 97%
4-(Trifluoromethoxy)benzamidoxime
Formula: C₈H₇F₃N₂O₂
MW: 220.15 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00085175
CAS Number: 56935-71-8