2-(Trifluoromethoxy)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Trifluoromethoxy)benzene-1-sulphonyl chloride
PC7436L-100G PC7436L-25G PC7436L-5G
APOSPC7436L-100GEA 101 EUR
APOSPC7436L-100G APOSPC7436L-25G APOSPC7436L-5G
2-(Trifluoromethoxy)benzenesulphonyl chloride 97%
2-(Trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 75…77 °C/11mmHg
Melting Pt: 27…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00052316
CAS Number: 103008-51-1
UN: 3261
ADR: 8,II