2-(Trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Warning
PC7436T-100G PC7436T-250G PC7436T-25G
APOSPC7436T-100GEA 106 EUR
APOSPC7436T-100G APOSPC7436T-250G APOSPC7436T-25G
2-(Trifluoromethoxy)benzoic acid 97%
2-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052325
CAS Number: 1979-29-9