2-(Trifluoromethoxy)benzoic acid hydrazide 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoromethoxy)benzhydrazide
PC7436U-1G PC7436U-5G PC7436U-25G
APOSPC7436U-1GEA 21.9 EUR
APOSPC7436U-1G APOSPC7436U-5G APOSPC7436U-25G
2-(Trifluoromethoxy)benzoic acid hydrazide 95%
2-(Trifluoromethoxy)benzoic acid hydrazide
Formula: C₈H₇F₃N₂O₂
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00204180
CAS Number: 175277-19-7