2-(Trifluoromethoxy)benzonitrile 97%
Supplier: Apollo Scientific
Danger
PC7438E-100G PC7438E-25G PC7438E-5G
APOSPC7438E-100GEA 95.7 EUR
APOSPC7438E-100G APOSPC7438E-25G APOSPC7438E-5G
2-(Trifluoromethoxy)benzonitrile 97%
2-(Trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 61…62 °C/8mmHg
Density: 1.245 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042409
CAS Number: 63968-85-4
EINECS: 264-584-0
UN: 3276
ADR: 6.1,III