4-(Trifluoromethoxy)benzyl alcohol 97%
Supplier: Apollo Scientific
Warning
PC7438SD-10G PC7438SD-25G PC7438SD-100G
APOSPC7438SD-10GEA 21.9 EUR
APOSPC7438SD-10G APOSPC7438SD-25G APOSPC7438SD-100G
4-(Trifluoromethoxy)benzyl alcohol 97%
4-(Trifluoromethoxy)benzyl alcohol
Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
Boiling Pt: 100…102 °C/12mmHg
Density: 1.326 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00036029
CAS Number: 1736-74-9