α-Bromo-3-(trifluoromethoxy)toluene 97%
Supplier: Apollo Scientific
Danger
Synonyms: m-(Trifluoromethoxy)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethoxy)benzene, 3-(Trifluoromethoxy)benzyl bromide
PC7438U-25G PC7438U-5G PC7438U-100G
APOSPC7438U-25GEA 132 EUR
APOSPC7438U-25G APOSPC7438U-5G APOSPC7438U-100G
α-Bromo-3-(trifluoromethoxy)toluene 97%
α-Bromo-e-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
Boiling Pt: 178…180 °C
Density: 1.572 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00061271
CAS Number: 159689-88-0
UN: 3265
ADR: 8,II