1-Bromo-4-(trifluoromethoxy)benzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethoxy)bromobenzene
PC7438W-250G PC7438W-100G PC7438W-1KG
APOSPC7438W-250GEA 46.4 EUR
APOSPC7438W-250G APOSPC7438W-100G APOSPC7438W-1KG
1-Bromo-4-(trifluoromethoxy)benzene 98%
1-Bromo-4-(trifluoromethoxy)benzene
Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Boiling Pt: 80 °C/50mmHg
Density: 1.622 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00040834
CAS Number: 407-14-7
EINECS: 206-979-2