1-Iodo-4-(trifluoromethoxy)benzene 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(Trifluoromethoxy)iodobenzene
PC7439G-100G
PC7439G-25G
APOSPC7439G-100GEA
42.2
EUR
APOSPC7439G-100G
APOSPC7439G-25G
1-Iodo-4-(trifluoromethoxy)benzene 97%
1-Iodo-4-(trifluoromethoxy)benzene
Formula:
C₇H₄F₃IO MW: 288.01 g/mol Boiling Pt: 72…74 °C/15mmHg Density: 1.909 g/cm³ (25 °C) Flash Pt: 77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042411 CAS Number: 103962-05-6 |