2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide
PC7439KM-100G PC7439KM-25G PC7439KM-5G
APOSPC7439KM-100GEA 227 EUR
APOSPC7439KM-100G APOSPC7439KM-25G APOSPC7439KM-5G
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone
Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
Melting Pt: 48…55 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052333
CAS Number: 103962-10-3
UN: 3261
ADR: 8 III