4-(Trifluoromethoxy)phenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: α,α,α-Trifluoro-4-hydroxyanisole
PC7439M-100G PC7439M-25G PC7439M-500G
APOSPC7439M-100GEA 55.1 EUR
APOSPC7439M-100G APOSPC7439M-25G APOSPC7439M-500G
4-(Trifluoromethoxy)phenol 98%
4-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
Boiling Pt: 80 °C/15mmHg
Melting Pt: 17…18 °C
Density: 1.375 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00040988
CAS Number: 828-27-3
EINECS: 212-583-0