4-(Trifluoromethoxy)phenyl isocyanate 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene
PC7439Q-1G PC7439Q-5G
APOSPC7439Q-1GEA 21.9 EUR
APOSPC7439Q-1G APOSPC7439Q-5G
4-(Trifluoromethoxy)phenyl isocyanate 98%
4-(Trifluoromethoxy)phenyl isocyanate
  • May form small amount of sediment.
Formula: C₈H₄F₃NO₂
MW: 203.12 g/mol
Boiling Pt: 78 °C/22mmHg
Melting Pt: –29 °C
Density: 1.346 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035702
CAS Number: 35037-73-1
UN: 2206
ADR: 6.1 III