4-(Trifluoromethoxy)thiophenol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethoxy)benzenethiol, 4-(Trifluoromethoxy)benzene-1-thiol
PC7439V-1G PC7439V-5G PC7439V-25G
APOSPC7439V-1GEA 45.1 EUR
APOSPC7439V-1G APOSPC7439V-5G APOSPC7439V-25G
4-(Trifluoromethoxy)thiophenol 95%
4-(Trifluoromethoxy)thiophenol
Formula: C₇H₅F₃OS
MW: 194.18 g/mol
Boiling Pt: 64…66 °C/15mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00052312
CAS Number: 169685-29-4
UN: 2810
ADR: 6.1,III