4-(Trifluoromethoxy)toluene 97%
Supplier: Apollo Scientific
Warning
PC7439W-25G PC7439W-5G PC7439W-100G
APOSPC7439W-25GEA 52.2 EUR
APOSPC7439W-25G APOSPC7439W-5G APOSPC7439W-100G
4-(Trifluoromethoxy)toluene 97%
4-(Trifluoromethoxy)toluene
Formula: C₈H₇F₃O
MW: 176.1400 g/mol
Boiling Pt: 134…135 °C
Density: 1.179 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00042413
CAS Number: 706-27-4
UN: 1993
ADR: 3,III