2-Bromo-6-fluorobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC7441-100G PC7441-25G PC7441-5G
APOSPC7441-100GEA 241 EUR
APOSPC7441-100G APOSPC7441-25G APOSPC7441-5G
2-Bromo-6-fluorobenzaldehyde 97%
2-Bromo-6-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
Boiling Pt: 100…102 °C/8mmHg
Melting Pt: 39…47 °C
Density: 1.7 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD03407341
CAS Number: 360575-28-6