5-Bromo-2-fluoro-4-methylsulphonyltoluene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-fluoro-4-methylphenyl methyl sulphone
PC7465-1G PC7465-500MG
APOSPC7465-1GEA 357 EUR
APOSPC7465-1G APOSPC7465-500MG
5-Bromo-2-fluoro-4-methylsulphonyltoluene
5-Bromo-2-fluoro-4-methylsulphonyltoluene
Formula: C₈H₈BrFO₂S
MW: 267.12 g/mol
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD09027746
CAS Number: 942474-79-5