4'-(Trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone
PC7490-100G PC7490-10G PC7490-25G
APOSPC7490-100GEA 72.5 EUR
APOSPC7490-100G APOSPC7490-10G APOSPC7490-25G
4'-(Trifluoromethyl)acetophenone
4'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 79…80 °C/8mmHg
Melting Pt: 30…33 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7