3-(Trifluoromethyl)anisole 98%
Supplier: Apollo Scientific
Warning
PC7496-100G PC7496-250G PC7496-25G
APOSPC7496-100GEA 58 EUR
APOSPC7496-100G APOSPC7496-250G APOSPC7496-25G
3-(Trifluoromethyl)anisole 98%
3-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 160 °C
Melting Pt: -65
Density: 1.217 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000385
CAS Number: 454-90-0
UN: 1993
ADR: 3 III