α,α,α-Trifluoro-2-tolualdehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoromethyl)benzaldehyde
PC7499-100G PC7499-25G PC7499-500G
APOSPC7499-100GEA 31.9 EUR
APOSPC7499-100G APOSPC7499-25G APOSPC7499-500G
α,α,α-Trifluoro-2-tolualdehyde 98%
α,α,α-Trifluoro-2-tolualdehyde
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 70…71 °C/16mmHg
Melting Pt: –40 °C
Density: 1.32 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00003337
CAS Number: 447-61-0
EINECS: 207-185-9