α,α,α-Trifluoro-3-tolualdehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23
PC7500-100G PC7500-25G PC7500-500G
APOSPC7500-100GEA 27.7 EUR
APOSPC7500-100G APOSPC7500-25G APOSPC7500-500G
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 83…86 °C/30mmHg
Density: 1.304 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1
UN: 3082
ADR: 9+EHS III