4-(Trifluoromethyl)benzamide 99%
Supplier: Apollo Scientific
Warning
PC7507-100G PC7507-25G PC7507-5G
APOSPC7507-100GEA 92.8 EUR
APOSPC7507-100G APOSPC7507-25G APOSPC7507-5G
4-(Trifluoromethyl)benzamide 99%
4-(Trifluoromethyl)benzamide
Formula: C₈H₆F₃NO
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00007998
CAS Number: 1891-90-3
EINECS: 217-571-9