3-(Trifluoromethyl)benzamidoxime 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N'-Hydroxy-3-(trifluoromethyl)-benzenecarboximidamide, N-Hydroxy-3-(trifluoromethyl)benzimidamide
PC7507L-100G
PC7507L-25G
PC7507L-5G
APOSPC7507L-100GEA
192
EUR
APOSPC7507L-100G
APOSPC7507L-25G
APOSPC7507L-5G
3-(Trifluoromethyl)benzamidoxime 97%
3-(Trifluoromethyl)benzamidoxime
Formula:
C₈H₇F₃N₂O MW: 204.15 g/mol Melting Pt: 85…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD02647312 CAS Number: 40067-80-9 |