2-(Trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Trifluoromethyl)benzene-1-sulphonyl chloride
PC7508-100G PC7508-25G PC7508-5G
APOSPC7508-100GEA 197 EUR
APOSPC7508-100G APOSPC7508-25G APOSPC7508-5G
2-(Trifluoromethyl)benzenesulphonyl chloride 98%
2-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 133…135 °C/14mmHg
Density: 1.585 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3265
ADR: 8 II