3-(Trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Trifluoromethyl)benzene-1-sulphonyl chloride
PC7509-100G PC7509-250G
APOSPC7509-100GEA 91.9 EUR
APOSPC7509-100G APOSPC7509-250G
3-(Trifluoromethyl)benzenesulphonyl chloride 98%
3-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 88…90 °C/6mmHg
Density: 1.526 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014724
CAS Number: 777-44-6
UN: 3265
ADR: 8 II