2-Chloro-3-(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-3-hydroxybenzotrifluoride
PC7543-1G PC7543-250MG
APOSPC7543-1GEA 59.6 EUR
APOSPC7543-1G APOSPC7543-250MG
2-Chloro-3-(trifluoromethyl)phenol 98%
2-Chloro-3-(trifluoromethyl)phenol
Formula: C₇H₄ClF₃O
Boiling Pt: 160 °C
Density: 1.479 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00213793
CAS Number: 138377-34-1
UN: 3265
ADR: 8,III