1,2-Dibromo-2-chloropentafluoropropane 97%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-1,2-dibromopentafluoropropane
PC7549-5G PC7549-1G
APOSPC7549-5GEA 611 EUR
APOSPC7549-5G APOSPC7549-1G
1,2-Dibromo-2-chloropentafluoropropane 97%
1,2-Dibromo-2-chloropentafluoropropane
Formula: C₃Br₂ClF₅
Boiling Pt: 116…117 °C
Storage Temperature: Ambient
CAS Number: 661-93-8