Methyl-3-amino-2-(trifluoromethyl)propionate
Supplier: Apollo Scientific
Danger
PC7593-1G PC7593-250MG PC7593-500MG
APOSPC7593-1GEA 0 EUR
APOSPC7593-1G APOSPC7593-250MG APOSPC7593-500MG
Methyl-3-amino-2-(trifluoromethyl)propionate
Methyl-3-amino-2-(trifluoromethyl)propionate
Formula: C₅H₈F₃NO₂
MW: 171.12 g/mol
Boiling Pt: 50 °C/0.2mmHg
Density: 1.345 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08457648
CAS Number: 914203-47-7