3-Bromo-4-fluorobenzamide
Supplier: Apollo Scientific
Warning
PC7598-250MG
APOSPC7598-250MGEA 21.9 EUR
APOSPC7598-250MG
3-Bromo-4-fluorobenzamide
3-Bromo-4-fluorobenzamide
Formula: C₇H₅BrFNO
MW: 218.0200 g/mol
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD03094431
CAS Number: 455-85-6