α,α,α-Trifluoro-m-toluoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride
PC7600-100G PC7600-25G PC7600-500G
APOSPC7600-100GEA 52.2 EUR
APOSPC7600-100G APOSPC7600-25G APOSPC7600-500G
α,α,α-Trifluoro-m-toluoyl chloride 98%
α,α,α-Trifluoro-m-toluoyl chloride
Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
Boiling Pt: 184…186 °C
Density: 1.38 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000680
CAS Number: 2251-65-2
EINECS: 218-844-5
UN: 3265
ADR: 8,II