α,α,α-Trifluoro-p-toluoyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-(Trifluoromethyl)benzoyl chloride, 4-(Trifluoromethyl)benzoylchloride, 4-(Trifluoromethyl)benzene-1-carbonyl chloride
PC7610-100G
PC7610-25G
APOSPC7610-100GEA
81.2
EUR
APOSPC7610-100G
APOSPC7610-25G
α,α,α-Trifluoro-p-toluoyl chloride 97%
α,α,α-Trifluoro-p-toluoyl chloride
Formula:
C₈H₄ClF₃O MW: 208.57 g/mol Boiling Pt: 188…190 °C Melting Pt: –3 °C Density: 1.404 g/cm³ (25 °C) Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000694 CAS Number: 329-15-7 EINECS: 206-342-9 UN: 3265 ADR: 8 II |