Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Supplier: Apollo Scientific
Synonyms: Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester, Ethyl 3-trifluoromethylpyrazole-4-carboxylate
PC7621-1G PC7621-5G
APOSPC7621-1GEA 193 EUR
APOSPC7621-1G APOSPC7621-5G
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Formula: C₇H₇F₃N₂O₂
MW: 208.14 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00052083
CAS Number: 155377-19-8