2-(Trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
Warning
PC7625-100G PC7625-25G PC7625-500G
APOSPC7625-100GEA 49.3 EUR
APOSPC7625-100G APOSPC7625-25G APOSPC7625-500G
2-(Trifluoromethyl)benzyl alcohol 97%
Formula: C₈H₇F₃O
Boiling Pt: 90 °C/20 mmHg
Density: 1.326 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00004621
CAS Number: 346-06-5
EINECS: 206-467-9