3-(Trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (3-(Trifluoromethyl)phenyl)methanol, 3-(Hydroxymethyl)benzotrifluoride
PC7626-100G PC7626-25G PC7626-5G
APOSPC7626-100GEA 64 EUR
APOSPC7626-100G APOSPC7626-25G APOSPC7626-5G
3-(Trifluoromethyl)benzyl alcohol 97%
3-(Trifluoromethyl)benzyl alcohol
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 79…80 °C/3mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00004645
CAS Number: 349-75-7
EINECS: 206-489-9