4-(Trifluoromethyl)benzyl alcohol 96%
Supplier: Apollo Scientific
Warning
Synonyms: (4-(Trifluoromethyl)phenyl)methanol, 4-(Hydroxymethyl)benzotrifluoride
PC7627-25G PC7627-5G PC7627-1G
APOSPC7627-25GEA 94.5 EUR
APOSPC7627-25G APOSPC7627-5G APOSPC7627-1G
4-(Trifluoromethyl)benzyl alcohol 96%
4-(Trifluoromethyl)benzyl alcohol
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 78…80 °C/4mmHg
Melting Pt: 18…20 °C
Density: 1.286 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004661
CAS Number: 349-95-1
EINECS: 206-494-6