4-(Trifluoromethyl)benzylamine 94%
Supplier: Apollo Scientific
Danger
PC7630-25G PC7630-100G
APOSPC7630-25GEA 31.9 EUR
APOSPC7630-25G APOSPC7630-100G
4-(Trifluoromethyl)benzylamine 94%
4-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 80…82 °C/15mmHg
Density: 1.229 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010220
CAS Number: 3300-51-4
EINECS: 221-971-9
UN: 2735
ADR: 8,III