3-(Trifluoromethyl)benzyl mercaptan, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Trifluoromethyl)benzylthiol
PC7634-10G PC7634-25G PC7634-1G
APOSPC7634-10GEA 102 EUR
APOSPC7634-10G APOSPC7634-25G APOSPC7634-1G
3-(Trifluoromethyl)benzyl mercaptan, Technical Grade
3-(Trifluoromethyl)benzyl mercaptan
Formula: C₈H₇F₃S
MW: 192.2 g/mol
Boiling Pt: 75…77 °C/5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042435
CAS Number: 25697-55-6
UN: 2810
ADR: 6.1 III