N-Methoxy-N-methyl-4-(trifluoromethyl)benzenecarboxamide
Supplier: Apollo Scientific
Warning
Synonyms: N-Methoxy-N-methyl-4-(trifluoromethyl)benzamide
PC7639-1G PC7639-5G
APOSPC7639-1GEA 125 EUR
APOSPC7639-1G APOSPC7639-5G
N-Methoxy-N-methyl-4-(trifluoromethyl)benzenecarboxamide
N-Methoxy-N-methyl-4-(trifluoromethyl)benzenecarboxamide
Formula: C₁₀H₁₀F₃NO₂
MW: 233.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689795
CAS Number: 116332-61-7