Methyl-3-amino-4-fluorobenzoate 99%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-amino-4-fluorobenzoate
PC7646-1G PC7646-25G PC7646-5G
APOSPC7646-1GEA 21.9 EUR
APOSPC7646-1G APOSPC7646-25G APOSPC7646-5G
Methyl-3-amino-4-fluorobenzoate 99%
Methyl 3-amino-4-fluorobenzoate
Formula: C₈H₈FNO₂
MW: 169.1500 g/mol
Melting Pt: 66…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689743
CAS Number: 369-26-6