N-Methyl-5-(trifluoromethyl)-2-pyridinamine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-Methyl-5-(trifluoromethyl)pyridin-2-amine, 2-(Methylamino)-5-(trifluoromethyl)pyridine
PC7647-1G
PC7647-5G
APOSPC7647-1GEA
68.3
EUR
APOSPC7647-1G
APOSPC7647-5G
N-Methyl-5-(trifluoromethyl)-2-pyridinamine 97%
N-Methyl-5-(trifluoromethyl)-2-pyridinamine
Formula:
C₇H₇F₃N₂ MW: 176.14 g/mol Melting Pt: 91…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689790 CAS Number: 937602-15-8 |