2,4,6-Trifluoro-N-methoxy-N-methylbenzenecarboxamide
Supplier: Apollo Scientific
Warning
Synonyms: N-Methoxy-N-methyl-2,4,6-trifluorobenzamide
PC7659-1G PC7659-500MG
APOSPC7659-1GEA 0 EUR
APOSPC7659-1G APOSPC7659-500MG
2,4,6-Trifluoro-N-methoxy-N-methylbenzenecarboxamide
2,4,6-Trifluoro-N-methoxy-N-methylbenzenecarboxamide
Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Melting Pt: 63…64 °C
Storage Temperature: Ambient
MDL Number: MFCD08689800
CAS Number: 937602-31-8