4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethyl)cyclohexane-1-carboxylic acid
PC7668D-1G PC7668D-5G PC7668D-25G
APOSPC7668D-1GEA 21.9 EUR
APOSPC7668D-1G APOSPC7668D-5G APOSPC7668D-25G
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture)
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₈H₁₁F₃O₂
MW: 196.17 g/mol
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00220049
CAS Number: 95233-30-0